Abnova
Manufacturer Code:MAB3347
Catalog # 89-111-364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 34B12 |
Immunogen | Native purified NOP58 from yeast nuclear protein preparations. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native NOP58.
For Research Use Only
Protein Aliases: NOP5; Nucleolar protein 5; Nucleolar protein 58; RNA-processing protein NOP58
Gene Aliases: NOP5; NOP58; O6108; YOR310C
UniProt ID: (Baker's yeast) Q12499
Entrez Gene ID: (Baker's yeast) 854487
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.