Invitrogen
Manufacturer Code:PA5114088
Catalog # PIPA5114088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NOP5/NOP58 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC120 MSSP Nap65 NOL5 NOP5 NOP5/NOP58 Nop58 NOP58 ribonucleoprotein NOP58 ribonucleoprotein homolog Nopp140 associated protein Nopp140-associated protein of 65 kDa Nucleolar protein 5 Nucleolar protein 58 nucleolar protein NOP5/NOP58 nucleolar protein NOP58 O6108 RNA-processing protein NOP58 SIK SIK similar protein SIK-similar protein YOR310C; NOP58 ribonucleoprotein homolog; Nucleolar protein 5; Nucleolar protein 58; nucleolar protein NOP5/NOP58
Gene Aliases: HSPC120; NOL5; NOP5; NOP5/NOP58; NOP58
UniProt ID: (Human) Q9Y2X3
Entrez Gene ID: (Human) 51602
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.