Invitrogen
Manufacturer Code:PA5100645
Catalog # PIPA5100645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NOP58 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC120 MSSP Nap65 Nol5 Nop5 NOP5/NOP58 Nop58 NOP58 ribonucleoprotein NOP58 ribonucleoprotein homolog Nopp140 associated protein nopp140-associated protein of 65 kDa Nucleolar protein 5 nucleolar protein 58 nucleolar protein NOP5/NOP58 nucleolar protein NOP58 O6108 RNA-processing protein NOP58 SIK SIK similar protein SIK-similar protein YOR310C; MSSP; NOP58 ribonucleoprotein homolog; Nopp140 associated protein; Nopp140-associated protein of 65 kDa; Nucleolar protein 5; Nucleolar protein 58; nucleolar protein NOP5/NOP58; nucleolar protein NOP58; SIK similar protein; SIK-similar protein
Gene Aliases: HSPC120; MSSP; Nap65; NOL5; NOP5; NOP5/NOP58; NOP58; SIK
UniProt ID: (Human) Q9Y2X3, (Mouse) Q6DFW4, (Rat) Q9QZ86
Entrez Gene ID: (Human) 51602, (Mouse) 55989, (Rat) 60373
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.