Invitrogen
Manufacturer Code:MA524641
Catalog # PIMA524641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2603 |
Immunogen | Recombit protein corresponding to Human NOP56 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NOP56 ribonucleoprotein homolog; NOP56 ribonucleoprotein homolog (yeast) NOP56 NOL5A SCA36; Nucleolar protein 56; Nucleolar protein 5A; nucleolar protein 5A (56kDa with KKE/D repeat)
Gene Aliases: NOL5A; NOP56; SCA36
UniProt ID: (Human) O00567
Entrez Gene ID: (Human) 10528
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.