Invitrogen
Manufacturer Code:PA541814
Catalog # PIPA541814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NOL5A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 56kDa with KKE/D repeat; NOP56 ribonucleoprotein homolog; NOP56 ribonucleoprotein homolog (yeast) NOP56 NOL5A SCA36; Nucleolar protein 56; Nucleolar protein 5A; nucleolar protein 5A (56kDa with KKE/D repeat)
Gene Aliases: 2310044F10Rik; NOL5A; NOP56; SCA36
UniProt ID: (Bovine) Q3SZ63, (Human) O00567, (Mouse) Q9D6Z1
Entrez Gene ID: (Rabbit) 100352750, (Horse) 100067054, (Bovine) 404165, (Dog) 612871, (Human) 10528, (Rat) 362214, (Guinea pig) 100717680, (Mouse) 67134
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.