Invitrogen
Manufacturer Code:PA559073
Catalog # PIPA559073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NOP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NOL1 NOP120 NSUN1 p120 NOL1/NOP2/Sun domain family member 1 nucleolar protein 1 120kDa nucleolar protein 2 homolog proliferating-cell nucleolar antigen p120 proliferation-associated nucleolar protein p120 putative ribosomal RNA methyltransferase NOP2; NOL1/NOP2/Sun domain family, member 1; NOP2 nucleolar protein homolog; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2 homolog; Probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; Proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2
Gene Aliases: 120kDa; A530002O17; NOL1; NOP120; NOP2; NSUN1; p120
UniProt ID: (Human) P46087
Entrez Gene ID: (Human) 4839, (Mouse) 110109, (Rat) 314969
Molecular Function:
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.