Abnova
Manufacturer Code:PAB27870
Catalog # 89-129-395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human NOP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant NOP2.
For Research Use Only
Protein Aliases: MGC117384; MGC149287; MGC149288; NOL1; NOL1/NOP2/Sun domain family, member 1; NOP120; NOP2 nucleolar protein homolog; NSUN1; Nucleolar protein 1; nucleolar protein 1, 120kDa; Nucleolar protein 2 homolog; p120; Probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; Proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2
Gene Aliases: NOL1; NOP120; NOP2; NSUN1; p120
UniProt ID: (Human) P46087
Entrez Gene ID: (Human) 4839
Molecular Function:
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.