Novus Biologicals
Manufacturer Code:NBP222227
Catalog # NBP222227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 750 and 800 of human Nucleolar Complex Protein 14 using the numbering given in entry NP_003694.1 (GeneID 8602). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C4orf9 chromosome 4 open reading frame 9 NOL14nucleolar protein 14 homolog (yeast) NOP14 homolog NOP14 nucleolar protein homolog (yeast) Nucleolar complex protein 14 nucleolar protein 14 nucleolar protein 14 homolog probable nucleolar complex protein 14 RES425 RES4-25; NOP14 nucleolar protein homolog; Nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14
Gene Aliases: C4orf9; NOL14; NOP14; RES4-25; RES425; UTP2
UniProt ID: (Human) P78316
Entrez Gene ID: (Human) 8602
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.