Invitrogen
Manufacturer Code:PA527408
Catalog # PIPA527408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 143 of Human p54/nrb |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 54 kDa nuclear RNA- and DNA-binding protein; 55 kDa nuclear protein; DNA-binding p52/p100 complex, 52 kDa subunit; NMT55; NMT55 NRB54 P54 P54NRB 54 kDa nuclear RNA- and DNA-binding protein 55 kDa nuclear protein DNA-binding p52/p100 complex 52 kDa subunit non-POU domain-containing octamer (ATGCAAAT) binding protein non-POU domain-containing octamer-binding protein p54(nrb); non-POU domain-containing octamer (ATGCAAAT) binding protein; Non-POU domain-containing octamer-binding protein; NonO protein; p54(nrb); p54nrb; protein phosphatase 1, regulatory subunit 114
Gene Aliases: MRXS34; NMT55; NONO; NRB54; P54; P54NRB; PPP1R114
UniProt ID: (Human) Q15233
Entrez Gene ID: (Human) 4841
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.