Invitrogen
Manufacturer Code:PA539315
Catalog # PIPA539315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human NOM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm1040 C7orf3; Nucleolar MIF4G domain-containing protein 1; protein phosphatase 1, regulatory subunit 113; SGD1 homolog
Gene Aliases: C7orf3; D5Kng1; Gm1040; NOM1; PPP1R113; SGD1
UniProt ID: (Human) Q5C9Z4, (Mouse) Q3UFM5
Entrez Gene ID: (Human) 64434, (Mouse) 433864
Molecular Function: RNA binding protein mRNA processing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.