Novus Biologicals
Manufacturer Code:NBP287936
Catalog # NB035558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human NOM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C7orf3 chromosome 7 open reading frame 3 FLJ16401 nucleolar MIF4G domain-containing protein 1 nucleolar protein with MIF4G domain 1 SGD1 SGD1 homolog; Nucleolar MIF4G domain-containing protein 1; protein phosphatase 1, regulatory subunit 113; SGD1 homolog
Gene Aliases: C7orf3; NOM1; PPP1R113; SGD1
UniProt ID: (Human) Q5C9Z4
Entrez Gene ID: (Human) 64434
Molecular Function: RNA binding protein mRNA processing factor nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.