Proteintech
Manufacturer Code:118151AP150UL
Catalog # 50-172-7477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NOLC1 Fusion Protein Ag2382 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; KIAA0035 NOLC1 NOPP130 NOPP140 NS5ATP13 Nucleolar 130 kDa protein Nucleolar phosphoprotein p130; Nopp140; Nucleolar 130 kDa protein; Nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphprotein 1; nucleolar phosphoprotein 140; Nucleolar phosphoprotein p130; nucleolar protein p130
Gene Aliases: KIAA0035; NOLC1; NOPP130; NOPP140; NS5ATP13; P130
UniProt ID: (Human) Q14978, (Rat) P41777, (Mouse) E9Q5C9
Entrez Gene ID: (Human) 9221, (Rat) 64896, (Mouse) 70769
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.