Invitrogen
Manufacturer Code:PA127677
Catalog # PIPA127677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human H/ACA ribonucleoprotein complex subunit 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: GAR1 homolog, ribonucleoprotein; GAR1 ribonucleoprotein homolog; H/ACA ribonucleoprotein complex subunit 1; NOLA1; Nucleolar protein family A member 1; nucleolar protein family A, member 1 (H/ACA small nucleolar RNPs); snoRNP protein GAR1
Gene Aliases: GAR1; NOLA1
UniProt ID: (Human) Q9NY12
Entrez Gene ID: (Human) 54433
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.