Novus Biologicals
Manufacturer Code:NB10075347
Catalog # NB10075347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody was a sequence from 1a.a. to 217a.a. from NOLA1(a 217a.a. long protein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: GAR1 homolog, ribonucleoprotein; GAR1 ribonucleoprotein homolog; GAR1 ribonucleoprotein homolog (yeast) H/ACA ribonucleoprotein complex subunit 1 NOLA1member 1 (H/ACA small nucleolar RNPs) Nucleolar protein family A member 1 snoRNP protein GAR1; H/ACA ribonucleoprotein complex subunit 1; Nucleolar protein family A member 1; nucleolar protein family A, member 1 (H/ACA small nucleolar RNPs); snoRNP protein GAR1
Gene Aliases: GAR1; NOLA1
UniProt ID: (Human) Q9NY12
Entrez Gene ID: (Human) 54433
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.