Invitrogen
Manufacturer Code:PA546234
Catalog # PIPA546234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NOL9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nucleolar protein 9; nucleolar protein 9 Grc3 NET6; Polynucleotide 5'-hydroxyl-kinase NOL9; polynucleotide 5'-kinase
Gene Aliases: 4632412I24Rik; 6030462G04Rik; AI449622; AW490720; Grc3; NET6; NOL9
UniProt ID: (Human) Q5SY16, (Mouse) Q8C8L5
Entrez Gene ID: (Human) 79707, (Dog) 479591, (Rabbit) 100353939, (Mouse) 74035, (Rat) 313744
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.