Invitrogen
Manufacturer Code:PA5115659
Catalog # PIPA5115659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NLRC5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI451557 AK220210 Caterpiller protein 16.1 CLR16.1 NLR family CARD domain containing 5 NLR family CARD domain containing 5 Nlrc5 NOD27 NOD4 NOD-like receptor C5 Nucleotide-binding oligomerization domain protein 27 Nucleotide-binding oligomerization domain protein 4 nucleotide-binding oligomerization domain leucine rich repeat and CARD domain containing 5 nucleotide-binding oligomerization domains 27 protein NLRC5; Caterpiller protein 16.1; CLR16.1; copine II; NLR family, CARD domain containing 5; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; Protein NLRC5
Gene Aliases: AI451557; AK220210; CLR16.1; Cpne2; NLRC5; NOD27; NOD4
UniProt ID: (Human) Q86WI3, (Mouse) C3VPR6
Entrez Gene ID: (Human) 84166, (Mouse) 434341, (Rat) 291861
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.