Novus Biologicals
Manufacturer Code:NB100524AF405
Catalog # NB100524W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D9 |
Immunogen | Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29] |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BLAU CARD15caspase recruitment domain protein 15 caspase recruitment domain family member 15 Caspase recruitment domain-containing protein 15 CDACUG CLR16.3 IBD1NLR family CARD domain containing 2 Inflammatory bowel disease protein 1 NLRC2 NOD-like receptor C2 nucleotide-binding oligomerization domain 2 nucleotide-binding oligomerization domain containing 2 nucleotide-binding oligomerization domain leucine rich repeat and CARD domaincontaining 2 nucleotide-binding oligomerization domain-containing protein 2 PSORAS1NOD2B; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NOD-like receptor C2; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2
Gene Aliases: ACUG; BLAU; CARD15; CD; CLR16.3; IBD1; NLRC2; NOD2; NOD2B; PSORAS1
UniProt ID: (Human) Q9HC29
Entrez Gene ID: (Human) 64127
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.