Abnova
Manufacturer Code:MAB12647
Catalog # 89-934-702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 26mNOD2 |
Immunogen | Mouse Nod2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against mouse Nod2.
For Research Use Only
Protein Aliases: ACUG BLAU CD Card15 F830032C23Rik IBD1 Nlrc2; caspase recruitment domain family, member 15; caspase recruitment domain-containing protein 15; nucleotide-binding oligomerization domain-containing protein 2
Gene Aliases: ACUG; BLAU; Card15; CD; F830032C23Rik; IBD1; Nlrc2
Entrez Gene ID: (Mouse) 257632
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.