Invitrogen
Manufacturer Code:PA5104317
Catalog # PIPA5104317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NOD2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACUG BLAU Card15 caspase recruitment domain family member 15 caspase recruitment domain protein 15 caspase recruitment domain-containing protein 15 CD CLR16.3 F830032C23Rik IBD1 Inflammatory bowel disease protein 1 NLR family CARD domain containing 2 NLRC2 NOD 2 NOD2 NOD-2 NOD2 protein NOD2B NOD-like receptor C2 nucleotide binding oligomerization domain containing 2 nucleotide-binding oligomerization domain 2 nucleotide-binding oligomerization domain containing 2 nucleotide-binding oligomerization domain leucine rich repeat and CARD domain containing 2 Nucleotide-binding oligomerization domain-containing protein 2 PSORAS1; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NOD-like receptor C2; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2
Gene Aliases: ACUG; BLAU; CARD15; CD; CLR16.3; F830032C23Rik; IBD1; NLRC2; NOD2; NOD2B; PSORAS1
UniProt ID: (Human) Q9HC29
Entrez Gene ID: (Human) 64127, (Mouse) 257632
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.