Novus Biologicals
Manufacturer Code:NB500253SS
Catalog # NB500253SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of human NOD2 (between residues 1000-1040). [UniProt# Q9HC29] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BLAU CARD15caspase recruitment domain protein 15 caspase recruitment domain family member 15 Caspase recruitment domain-containing protein 15 CDACUG CLR16.3 IBD1NLR family CARD domain containing 2 Inflammatory bowel disease protein 1 NLRC2 NOD-like receptor C2 nucleotide-binding oligomerization domain 2 nucleotide-binding oligomerization domain containing 2 nucleotide-binding oligomerization domain leucine rich repeat and CARD domaincontaining 2 nucleotide-binding oligomerization domain-containing protein 2 PSORAS1NOD2B; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NOD-like receptor C2; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2
Gene Aliases: ACUG; BLAU; CARD15; CD; CLR16.3; IBD1; NLRC2; NOD2; NOD2B; PSORAS1
UniProt ID: (Human) Q9HC29
Entrez Gene ID: (Human) 64127
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.