Abnova
Manufacturer Code:H00064127D01P
Catalog # 89-002-998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NOD2 (AAI56572.1 1 a.a. ~ 1040 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human NOD2 protein.
For Research Use Only
Protein Aliases: ACUG; BLAU; CARD15; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; CD; CLR16.3; IBD1; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NLRC2; NOD-like receptor C2; NOD2B; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2; PSORAS1
Gene Aliases: ACUG; BLAU; CARD15; CD; CLR16.3; IBD1; NLRC2; NOD2; NOD2B; PSORAS1
UniProt ID: (Human) Q9HC29
Entrez Gene ID: (Human) 64127
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.