Abnova
Manufacturer Code:PAB19044
Catalog # 89-120-599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at C-terminus of human NOD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of NOD1.
For Research Use Only
Protein Aliases: CARD4; caspase recruitment domain family, member 4; Caspase recruitment domain-containing protein 4; CLR7.1; NLR family, CARD domain containing 1; NLRC1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; Nucleotide-binding oligomerization domain-containing protein 1
Gene Aliases: CARD4; CLR7.1; NLRC1; NOD1
UniProt ID: (Human) Q9Y239
Entrez Gene ID: (Human) 10392
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.