Invitrogen
Manufacturer Code:PA141264
Catalog # 01-675-610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to residues 87-103 and 231-246 of human INHAT-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp564C186 FLJ35172 NET15 NET7 NIR; NOC2-like nucleolar associated transcriptional repressor; NOC2-like protein; NOC2L nucleolar associated transcriptional repressor; novel INHAT (inhibitor of histone acetyltransferase) repressor; Novel INHAT repressor; nucleolar complex associated 2 homolog; Nucleolar complex protein 2 homolog; Protein NOC2 homolog; protein phosphatase 1, regulatory subunit 12
Gene Aliases: NET15; NET7; NIR; NOC2L; PPP1R112
UniProt ID: (Human) Q9Y3T9, (Bovine) Q3SYU1
Entrez Gene ID: (Human) 26155, (Bovine) 508638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.