OriGene
Manufacturer Code:TA808806
Catalog # 50-168-2381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-234 of human NOB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adenocarcinoma antigen recognized by T lymphocytes 4; ART-4 MST158 MSTP158 NOB1P PSMD8BP1; nin one binding protein; NIN1/RPN12 binding protein 1 homolog; Phosphorylation regulatory protein HP-10; Protein ART-4; PSMD8 binding protein 1; RNA-binding protein NOB1
Gene Aliases: ART-4; ART4; MST158; MSTP158; NOB1; NOB1P; PSMD8BP1
UniProt ID: (Human) Q9ULX3
Entrez Gene ID: (Human) 28987
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.