Abnova
Manufacturer Code:PAB15596
Catalog # 89-117-670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to <i>Saccharomyces cerevisiae</i> NOB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant NOB1.
For Research Use Only
Protein Aliases: 20S-pre-rRNA D-site endonuclease NOB1; NIN1-binding protein; Pre-rRNA-processing endonuclease NOB1; rRNA-binding endoribonuclease; YOR29-07
Gene Aliases: NOB1; YOR056C; YOR29-07
UniProt ID: (Baker's yeast) Q08444
Entrez Gene ID: (Baker's yeast) 854221
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.