Proteintech
Manufacturer Code:100912AP150UL
Catalog # 50-172-6016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NOB1 Fusion Protein Ag0141 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenocarcinoma antigen recognized by T lymphocytes 4; ART 4 ART4 MST158 MSTP158 NOB1 NOB1P Protein ART 4 PSMD8BP1 RNA binding protein NOB1; nin one binding protein; NIN1/RPN12 binding protein 1 homolog; Phosphorylation regulatory protein HP-10; Protein ART-4; PSMD8 binding protein 1; RNA-binding protein NOB1
Gene Aliases: 1700021I09Rik; ART-4; ART4; MST158; MSTP158; NOB1; NOB1P; PSMD8BP1
UniProt ID: (Human) Q9ULX3, (Rat) Q6VEU1, (Mouse) Q8BW10
Entrez Gene ID: (Human) 28987, (Rat) 291996, (Mouse) 67619
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.