Invitrogen
Manufacturer Code:PA567597
Catalog # PIPA567597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human NO66. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L8 histidine hydroxylase; Bifunctional lysine-specific demethylase and histidyl-hydroxylase NO66; Histone lysine demethylase NO66; hsNO66; lysine-specific demethylase NO66; MAPJD NO66 MYC-associated protein with JmjC domain hsNO66 lysine-specific demethylase NO66 nucleolar protein 66; myc associated protein with JmjC domain; MYC-associated protein with JmjC domain; Nucleolar protein 66; Ribosomal oxygenase 1; Ribosomal oxygenase NO66; ROX; up-regulated in lung cancer 2
Gene Aliases: C14orf169; hsNO66; MAPJD; MNCb-7109; NO66; RIOX1; ROX; URLC2
UniProt ID: (Mouse) Q9JJF3, (Human) Q9H6W3, (Rat) D3ZU57
Entrez Gene ID: (Mouse) 71952, (Human) 79697, (Rat) 314300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.