Proteintech
Manufacturer Code:134422AP150UL
Catalog # 50-172-8747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NNT Fusion Protein Ag4259 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NAD(P) transhydrogenase, mitochondrial; Nicotinamide nucleotide transhydrogenase; Nicotinamide nucleotide transhydrogenase (NAD(P)+ transhydrogenase); NNT; Pyridine nucleotide transhydrogenase
Gene Aliases: 4930423F13Rik; AI323702; BB168308; GCCD4; NNT
UniProt ID: (Human) Q13423
Entrez Gene ID: (Human) 23530, (Rat) 310378, (Mouse) 18115
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.