Invitrogen
Manufacturer Code:PA533972
Catalog # PIPA533972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human NMUR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; Neuromedin-U receptor 1; NMU-R1; NMUR1 G protein-coupled receptor 66 GPR66 FM-3 FM3 G-protein coupled receptor 66 GPC-R Neuromedin-U receptor 1 NMU-R1 NMU1R Neuromedin U receptor 1
Gene Aliases: (FM-3); FM-3; FM3; GPC-R; GPR66; NMU1R; NMUR1
UniProt ID: (Human) Q9HB89
Entrez Gene ID: (Human) 10316
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.