Invitrogen
Manufacturer Code:PA5113461
Catalog # PIPA5113461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Neuromedin-U receptor 1 protein (360-426AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (FM-3) FM3 FM-3 G protein-coupled receptor 66 G protein-coupled receptor FM-3 GPC-R GPR66 G-protein coupled receptor 66 G-protein coupled receptor FM-3 neuromedin U receptor 1 Neuromedin-U receptor 1 NMU1R NmU-R NMUR1 NMU-R1; G protein-coupled receptor 66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; Neuromedin-U receptor 1; NMU-R1
Gene Aliases: (FM-3); FM-3; FM3; GPC-R; GPR66; NmU-R; NMU-R1; NMU1R; NMUR1
UniProt ID: (Human) Q9HB89, (Mouse) O55040
Entrez Gene ID: (Human) 10316, (Mouse) 14767
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.