Invitrogen
Manufacturer Code:PA579746
Catalog # PIPA579746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human NMU. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; Neuromedin-U; Neuromedin-U Neuromedin-U-25 NmU-25 NMU; Neuromedin-U-23; Neuromedin-U-25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU
Gene Aliases: NMU
UniProt ID: (Human) P48645, (Rat) P12760, (Mouse) Q9QXK8
Entrez Gene ID: (Human) 10874, (Rat) 63887, (Mouse) 56183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.