Novus Biologicals
Manufacturer Code:NBP232244
Catalog # NBP232244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 100 to 150 of human N-myristoyltransferase 2 using the numbering given in entry NP_004799.1 (GeneID 9397). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.97 glycylpeptide N-tetradecanoyltransferase 2 Myristoyl-CoA:protein N-myristoyltransferase 2 NMT 2 N-myristoyltransferase 2 Peptide N-myristoyltransferase 2 Type II N-myristoyltransferase; Glycylpeptide N-tetradecanoyltransferase 2; glycylpeptide N-tetradecanoyltransferase 2 variant 3; glycylpeptide N-tetradecanoyltransferase 2 variant 4; Myristoyl-CoA:protein N-myristoyltransferase 2; NMT 2; Peptide N-myristoyltransferase 2; Type II N-myristoyltransferase
Gene Aliases: NMT2
UniProt ID: (Human) O60551
Entrez Gene ID: (Human) 9397
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.