Invitrogen
Manufacturer Code:PA578082
Catalog # PIPA578082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human NMT1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative, short form NMT-S; Glycylpeptide N-tetradecanoyltransferase 1; long form, NMT-L; Myristoyl-CoA:protein N-myristoyltransferase 1; N-myristoyltransferase 1 NMT NMT1 NMT1; NMT 1; Peptide N-myristoyltransferase 1; type I N-myristoyltransferase
Gene Aliases: AW536594; NMT; NMT1
UniProt ID: (Human) P30419, (Mouse) O70310, (Rat) Q8K1Q0
Entrez Gene ID: (Human) 4836, (Mouse) 18107, (Rat) 259274
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.