Invitrogen
Manufacturer Code:PA5113202
Catalog # PIPA5113202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3 protein (1-215AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933408N02Rik FKSG76 naMN adenylyltransferase 1 NaMN adenylyltransferase 3 nicotinamide mononucleotide adenylyltransferase 3 nicotinamide nucleotide adenylyltransferase 3 Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3 Nicotinamide-nucleotide adenylyltransferase 3 nicotinate-nucleotide adenylyltransferase 1 Nicotinate-nucleotide adenylyltransferase 3 NMN adenylyltransferase 3 NMN/NaMN adenylyltransferase 3 NMNAT3 PNAT3 PNAT-3 Pyridine nucleotide adenylyltransferase 3; naMN adenylyltransferase 1; NaMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; Nicotinamide-nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; nicotinate-nucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3; PNAT-3; Pyridine nucleotide adenylyltransferase 3
Gene Aliases: 4933408N02Rik; FKSG76; NMNAT3; PNAT-3; PNAT3
UniProt ID: (Human) Q96T66, (Mouse) Q99JR6
Entrez Gene ID: (Human) 349565, (Mouse) 74080
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.