Abnova
Manufacturer Code:PAB27576
Catalog # 89-129-087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 149-162 at internal region of human NMNAT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of NMNAT3.
For Research Use Only
Protein Aliases: NaMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; Nicotinamide-nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; Nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3; PNAT-3; PNAT3; Pyridine nucleotide adenylyltransferase 3
Gene Aliases: FKSG76; NMNAT3; PNAT-3; PNAT3
UniProt ID: (Human) Q96T66
Entrez Gene ID: (Human) 349565
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.