Abnova
Manufacturer Code:MAB9716
Catalog # 89-113-738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AT4A2 |
Immunogen | Recombinant protein corresponding to amino acids 1-279 of human NMNAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant NMNAT1.
For Research Use Only
Protein Aliases: NaMN adenylyltransferase 1; nicotinamide mononucleotide adenylyltransferase 1; Nicotinamide-nucleotide adenylyltransferase 1; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; NMNAT; PNAT-1; PNAT1; pyridine nucleotide adenylyltransferase 1
Gene Aliases: LCA9; NMNAT; NMNAT1; PNAT1
UniProt ID: (Human) Q9HAN9
Entrez Gene ID: (Human) 64802
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.