Invitrogen
Manufacturer Code:PA543246
Catalog # PIPA543246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NMNAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deamido-NAD(+) diphosphorylase; deamido-NAD(+) pyrophosphorylase; NAD(+) diphosphorylase; NAD(+) pyrophosphorylase; NaMN adenylyltransferase 1; nicotinamide mononucleotide adenylyl transferase; nicotinamide mononucleotide adenylyl transferase 1; Nicotinamide mononucleotide adenylyltransferase 1; nicotinamide mononucleotide adenylyltransferase 1-like protein; Nicotinamide nucleotide adenylyltransferase 1 NMNAT PNAT-1 PNAT1; Nicotinamide-nucleotide adenylyltransferase 1; nicotinamide/nicotinate mononucleotide adenylyltransferase 1; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; pyridine nucleotide adenylyltransferase 1
Gene Aliases: 2610529L11Rik; 5730441G13Rik; D4Cole1e; id:ibd5068; im:7144541; LCA9; NMNAT; NMNAT1; PNAT1; zgc:110243
UniProt ID: (Human) Q9HAN9, (Bovine) Q0VD50, (Mouse) Q9EPA7
Entrez Gene ID: (Human) 64802, (Rabbit) 100356468, (Dog) 100856724, (Goat) 102172286, (Horse) 100057371, (Bovine) 522863, (Rat) 298653, (Guinea pig) 100727955, (Mouse) 66454, (Zebrafish) 550322
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.