Proteintech
Manufacturer Code:113991AP150UL
Catalog # 50-172-7125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NMNAT1 Fusion Protein Ag1944 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deamido-NAD(+) diphosphorylase; deamido-NAD(+) pyrophosphorylase; NAD(+) diphosphorylase; NAD(+) pyrophosphorylase; NaMN adenylyltransferase 1; NaMN adenylyltransferase 1 NMNAT1 PNAT 1 PNAT1; nicotinamide mononucleotide adenylyl transferase; Nicotinamide mononucleotide adenylyltransferase 1; Nicotinamide-nucleotide adenylyltransferase 1; nicotinamide/nicotinate mononucleotide adenylyltransferase 1; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; pyridine nucleotide adenylyltransferase 1
Gene Aliases: 2610529L11Rik; 5730441G13Rik; D4Cole1e; LCA9; NMNAT; NMNAT1; PNAT1
UniProt ID: (Human) Q9HAN9, (Mouse) Q9EPA7
Entrez Gene ID: (Human) 64802, (Pig) 102160928, (Mouse) 66454
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.