Novus Biologicals
Manufacturer Code:NBP29469320UL
Catalog # NB023911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-302 of human NMNAT2 (NP_733820.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf15 chromosome 1 open reading frame 15 EC 2.7.7.1 EC 2.7.7.18 KIAA0479MGC2756 NaMN adenylyltransferase 2 nicotinamide mononucleotide adenylyltransferase 2 nicotinamide nucleotide adenylyltransferase 2 Nicotinate-nucleotide adenylyltransferase 2 NMN adenylyltransferase 2 PNAT2 pyridine nucleotide adenylyltransferase 2; NaMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; Nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; pyridine nucleotide adenylyltransferase 2
Gene Aliases: C1orf15; KIAA0479; NMNAT2; PNAT2
UniProt ID: (Human) Q9BZQ4
Entrez Gene ID: (Human) 23057
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.