Invitrogen
Manufacturer Code:MA520379
Catalog # PIMA520379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F5 |
Immunogen | NME5 (NP&lowbar003542 121 a.a. &bsim 212 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; nm23-H5; NM23-H5 NM23H5 RSPH23 IPIA-beta NDK-H 5 NDP kinase homolog 5 inhibitor of p53-induced apoptosis-beta nucleoside diphosphate kinase homolog 5 radial spoke 23 homolog testis-specific nm23 homolog; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); Nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; Testis-specific nm23 homolog
Gene Aliases: NM23-H5; NM23H5; NME5; RSPH23
UniProt ID: (Human) P56597
Entrez Gene ID: (Human) 8382
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.