Invitrogen
Manufacturer Code:PA521442
Catalog # PIPA521442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 212 of NME5 (Uniprot IDNo.P56597) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: expressed in non-metastatic cells 5; Inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; nm23-H5; NM23-H5 NM23H5 RSPH23 IPIA-beta NDK-H 5 NDP kinase homolog 5 inhibitor of p53-induced apoptosis-beta nucleoside diphosphate kinase homolog 5 radial spoke 23 homolog testis-specific nm23 homolog; nm23-M5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); Nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; Rsph23; Testis-specific nm23 homolog
Gene Aliases: 1700019D05Rik; NM23-H5; Nm23-M5; NM23H5; NME5; RSPH23
UniProt ID: (Human) P56597, (Mouse) Q99MH5
Entrez Gene ID: (Human) 8382, (Mouse) 75533
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.