OriGene
Manufacturer Code:CF501110
Catalog # 50-166-0147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Full length human recombit protein of human NME4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NDK; NDP kinase D; NDP kinase, mitochondrial; NDPK-D NM23H4 nm23-H4; NDPKD; nm23-H4; non-metastatic cells 4, protein expressed in; Nucleoside diphosphate kinase D; Nucleoside diphosphate kinase, mitochondrial
Gene Aliases: NDPK-D; nm23-H4; NM23D; NM23H4; NME4
UniProt ID: (Human) O00746
Entrez Gene ID: (Human) 4833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.