Invitrogen
Manufacturer Code:MA535687
Catalog # PIMA535687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1412 |
Immunogen | A synthesized peptide derived from human NME2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2.7.13.3 C-myc purine-binding transcription factor PUF Histidine protein kinase NDKB NDK B NDKB NDPKB NDPK-B nm23-2 NM23B nm23-H2 nm23-M2 Nme2 Nucleoside diphosphate kinase B P18 p18-12d PUF; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; Histidine protein kinase NDKB; NDK B; NDP kinase alpha; NDP kinase B; nm23-M2; non-metastatic cells 2, protein (NM23B) expressed in; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; P18
Gene Aliases: NDKB; nm23-2; NM23-H2; nm23-M2; NM23B; Nme2; p18-12d
UniProt ID: (Rat) P19804, (Mouse) Q01768
Entrez Gene ID: (Rat) 83782, (Mouse) 18103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.