Invitrogen
Manufacturer Code:PA599178
Catalog # PIPA599178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NM23 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2.7.13.3 c-myc purine-binding transcription factor PUF c-myc transcription factor epididymis secretory sperm binding protein Li 155an expressed in non-metastatic cells 2 expressed in non-metastatic cells 2 protein HEL-S-155an histidine protein kinase NDKB NDK B NDKB NDP kinase alpha NDP kinase B NDPKB NDPK-B nm23-2 NM23B nm23-H2 nm23-M2 NME/NM23 nucleoside diphosphate kinase 2 Nme2 non-metastatic cells 2 protein (NM23) expressed in non-metastatic cells 2 protein (NM23B) expressed in nucleoside diphosphate kinase B nucleoside-diphosphate kinase 2 nucleotide diphosphate kinase B P18 p18-12d PUF; C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; Histidine protein kinase NDKB; NDK B; NDP kinase alpha; NDP kinase B; nm23-H2; nm23-M2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18
Gene Aliases: NDKB; NDPK-B; NDPKB; nm23-2; NM23-H2; nm23-M2; NM23B; NME2; p18-12d; PUF
UniProt ID: (Human) P22392, (Rat) P19804, (Mouse) Q01768
Entrez Gene ID: (Human) 4831, (Rat) 83782, (Mouse) 18103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.