Invitrogen
Manufacturer Code:PA527488
Catalog # PIPA527488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 105 of Human nm23-H2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2 protein; HEL-S-155an; Histidine protein kinase NDKB; NDK B; NDKB NDPK-B NDPKB NM23-H2 NM23B PUF NDP kinase B c-myc purine-binding transcription factor PUF c-myc transcription factor histidine protein kinase NDKB non-metastatic cells 2 protein (NM23) expressed in nucleoside diphosphate kinase B nucleotide diphosphate kinase B; NDP kinase B; nm23-H2; nm23-M2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18
Gene Aliases: NDKB; NDPK-B; NDPKB; NM23-H2; nm23-M2; NM23B; NME2; PUF
UniProt ID: (Human) P22392, (Mouse) Q01768
Entrez Gene ID: (Human) 4831, (Mouse) 18103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.