Invitrogen
Manufacturer Code:PA589022
Catalog # PIPA589022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-267 of human NME1-NME2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; GAAD; Granzyme A-activated DNase; HEL-S-155an; Histidine protein kinase NDKB; Metastasis inhibition factor NM23; NDK A; NDK B; NDP kinase A; NDP kinase alpha; NDP kinase B; NDP kinase beta; NDPK-A; NM23-H1; nm23-H2; nm23-M1; nm23-M2; non-metastatic cells 1, protein (NM23A) expressed in; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; Nucleoside diphosphate kinase A; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase; nucleotide diphosphate kinase B; nucloside diphosphate kinase; P18; Tumor metastatic process-associated protein
Gene Aliases: AL024257; AWD; GAAD; NB; NBS; NDKA; NDKB; NDPK-A; NDPK-B; NDPKA; NDPKB; NM23; nm23-2; NM23-H1; NM23-H2; NM23-M1; nm23-M2; NM23A; NM23B; NME1; NME2; p18-12d; PUF
UniProt ID: (Human) P15531, (Human) P22392, (Mouse) P15532, (Rat) Q05982, (Rat) P19804, (Mouse) Q01768
Entrez Gene ID: (Human) 4830, (Human) 4831, (Mouse) 18102, (Rat) 191575, (Rat) 83782, (Mouse) 18103
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.