OriGene
Manufacturer Code:UM800024CF
Catalog # 50-168-5181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB93 |
Immunogen | Full length human recombit protein of human NME1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AWD GAAD NB NBS NDKA NDPK-A NDPKA NM23 NM23-H1; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); GAAD; Granzyme A-activated DNase; Metastasis inhibition factor NM23; NDK A; NDP kinase A; NDP kinase beta; nm23-C1; NM23-H1; non-metastatic cells 1, protein (NM23A) expressed in; Nucleoside diphosphate kinase A; nucloside diphosphate kinase; Tumor metastatic process-associated protein
Gene Aliases: AWD; GAAD; NB; NBS; NDKA; NDPK-A; NDPKA; NM23; NM23-C1; NM23-H1; NM23A; NME1
UniProt ID: (Human) P15531, (Dog) Q50KA9, (Rat) Q05982
Entrez Gene ID: (Human) 4830, (Dog) 480558, (Rat) 191575
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.