Invitrogen
Manufacturer Code:PA571669
Catalog # PIPA571669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 904-933 amino acids from the C-terminal region of human GRIN3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN3B; glutamate [NMDA] receptor subunit 3B; Glutamate receptor ionotropic, NMDA 3B; glutamate receptor, ionotropic, N-methyl-D-aspartate 3B; GRIN3B glutamate receptor ionotropic N-methyl-D-aspartate 3B NR3B NMDA receptor subunit 3B NMDA type glutamate receptor subunit NR3B; N-methyl-D-aspartate receptor subtype 3B; NMDA receptor subunit 3B; NMDA type glutamate receptor subunit NR3B; NMDAR3B
Gene Aliases: GluN3B; GRIN3B; NR3B
UniProt ID: (Human) O60391
Entrez Gene ID: (Human) 116444
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.