Invitrogen
Manufacturer Code:PA519347
Catalog # PIPA519347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EAPPHSGRPGSQ) corresponding to the C-terminus amino acids of GRIN3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN3B; glutamate [NMDA] receptor subunit 3B; Glutamate receptor ionotropic, NMDA 3B; glutamate receptor, ionotropic, N-methyl-D-aspartate 3B; GRIN3B glutamate receptor ionotropic N-methyl-D-aspartate 3B NR3B NMDA receptor subunit 3B NMDA type glutamate receptor subunit NR3B NMDAR; N-methyl-D-aspartate receptor subtype 3B; NMDA receptor subunit 3B; NMDA type glutamate receptor subunit NR3B; NMDAR3B
Gene Aliases: GluN3B; GRIN3B; NR3B
UniProt ID: (Human) O60391
Entrez Gene ID: (Human) 116444
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.