Novus Biologicals
Manufacturer Code:NBP261746
Catalog # NBP2-61746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6E9A8 |
Immunogen | Purified recombinant fragment of human GRIN2B (AA: extra 27-163) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: GluN2B; glutamate &lsqbNMDA] receptor subunit epsilon-2 glutamate receptor subunit epsilon-2 glutamate receptor ionotropic N-methyl D-aspartate 2B hNR3 MGC142180 NMDAR2BMGC142178 N-methyl D-aspartate receptor subtype 2B N-methyl-D-aspartate receptor subunit 3 NR2B NR3; Glutamate [NMDA] receptor subunit epsilon-2; Glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, N-methyl D-aspartate 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3; NMDAR2B; NR3
Gene Aliases: EIEE27; GluN2B; GRIN2B; hNR3; MRD6; NMDAR2B; NR2B
UniProt ID: (Human) Q13225
Entrez Gene ID: (Human) 2904
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.